Temperature indicating strips
Supplier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0646.0072 0646.0073 0646.0074 0646.0075 0646.0076 0646.0077
620-9122EA 32.8 GBP
620-9122 620-9123 620-9124 620-9125 620-9126 620-9127
Temperature indicating strips
Temperature Indicating Strips
These indicators are self-adhesive, temperature proof foils with temperature sensitive elements for temperature control and regulation. They are particularly suitable for monitoring temperature in small objects.
  • Change in colour within two seconds
  • Practical booklet with 10 indicators
  • Maximum storage temperature: 25 °C; storage in a refrigerator is recommended