Pocket thermo-hygrometers, testo 606
Supplier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0560.6060 0560.6062
620-3309EA 98.3 GBP
620-3309 620-3310
Pocket thermo-hygrometers, testo 606
Hygrometers Digital Thermo-Hygrometers
These testo thermo-hygrometers are convenient and easy-to operate pocket-sized material moisture measuring instrument. It is used for the measurement of wood moisture, characteristic curves are available for beech, spruce, larch, oak, pine and maple. For locating wet spots in building materials, curves are additionally stored for cement screed, plaster, anhydrite screed, cement mortar, lime mortar and bricks.
  • Precise measurement of wood moisture
  • Characteristics curves for locating wet spots in building materials
  • Hold-function for convenient read-out of
More Product Information