Digital thermometers, KM 330 and KM 340
Supplier: COMARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KM330 KM340
620-3161EA 217 GBP
620-3161 620-1174
Digital thermometers, KM 330 and KM 340
Thermometers Digital Thermometers
These multi-purpose thermometers are delivered in a protective rubber boot.
  • Last measurement storage
  • Maximum temperature memory
  • Type KM 340 with twin inputs for direct or differential measurements between two thermocouples
Type K thermocouple probes.