tempmate.®-S2 Single Use Temperature and Humidity Data Logger
Supplier: tempmate
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
S2H-000
620-2828EA 641 GBP
620-2828
tempmate.®-S2 Single Use Temperature and Humidity Data Logger
Data Loggers
Outstanding single-use temperature and humidity logger, for reliable cold chain monitoring. Integrated USB port and PDF and CSV generating software on-board.
  • Cost-effective and reliable
  • Smart design, intuitive operation
  • Waterproof and food grade packing
  • No additional hardware or software required
  • Detailed PDF and CSV report including graph and table
  • Intelligent LEDs for status monitoring
  • Extended shelf-life of two years
tempmate.®-S2 loggers are specially designed for temperature and/or humidity monitoring in any cold chain logistics. As a pre-More Product Information
Order Now
SPECIFICATIONS
Model Single-use data logger, temperature/relative humidity
Sensor Ambient light sensor
Environmentally Preferable
Interface USB
Protection class IP 52
Weight 30 g
Data memory - No. of readings 32000 values (T/rH)
Shelf life 2 years
Batteries CR2450 lithium metal button cell battery