Digital oil tester, testo 270
Supplier: Testo
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0563.2750
620-2783EA 402 GBP
620-2783
Digital oil tester, testo 270
Thermometers Digital Thermometers
This digital thermometer ensures cooking oil quality in restaurant and food service environments.
  • TPM from 0,5 to 40% TPM to indicate whether oil must be changed
  • Visible three-colour bar and audible alarm
  • Limit values to optimise cooking oil use
  • Reliable, robust and easy to clean
  • No requirement for sensor to cool down between measurements
Capacitive TPM (total polar materials) sensor in metal and breakage-proof.
Delivery information: Supplied with a dishwasher-proof TopSafe protective case, hand strap, reference oil, aluminium carrying case and AAA batteries.