Handheld digital microscopes, Dino-Lite Universal
Supplier: Dino-Lite / IDCP B.V.
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AM4113T AM4113ZT AM4013MZT AM4013MT AM4115T AM4115ZTW AM4515ZT AM4815ZT AM4115ZT AM7115MZT AM7515MZT AM7915MZT AM7115MZTW AM4515T AM7515MT2A
630-1157EA 467 GBP
630-1157 630-1165 630-1345 620-2617 620-2618 620-2620- 620-2621 620-2622 620-2619 630-2656 630-2658 630-2752 IDCPAM7115MZTW IDCPAM4515T IDCPAM7515MT2A
Handheld digital microscopes, Dino-Lite Universal
Microscopes
The Universal series provides a broad range of products with high image quality to satisfy the most demanding user. Magnification range: 10 to 220×.
  • High speed USB 2.0 connection
  • Metal or composite housing
  • Edge series models with high quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, Smart phone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on reflective surfaces (some models only)