VWR® Traceable® Ultra, Triple-Display Thermometers
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
620-2334EA 102 GBP
620-2334 620-2335 620-2336 620-2337 620-2338 620-2339 620-2340
VWR® Traceable® Ultra, Triple-Display Thermometers
Thermometers Digital Thermometers
Digital thermometers are designed to accurately monitor the temperature of fridges, freezers, incubators, water baths or during transport, in one or two different areas with enclosed temperature-buffered sensors or bullet sensors.
  • Smart alarm: Visual and audible alarm even if the unit returns to non alarm conditions
  • Triple-display simultaneously shows maximum/minimum and current temperatures
  • Maximum/minimum indicator
Probes: 1 or 2 bullet sensors/1 or 2 bottle probes with non toxic glycol, in 2 versions; 5 ml bottle or micro bottle to simulateMore Product Information