VWR® Traceable®, Waterproof Thermometers
Supplier: VWR Collection
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
620-2092EA 75.5 GBP
620-2092 620-2099
VWR® Traceable®, Waterproof Thermometers
Thermometers Digital Thermometers
This waterproof and shockproof thermometer is a safe replacement for any hazardous mercury instrument. The probe is resistant to most chemicals.
  • Waterproof construction for wet labs, wash-down areas and all field applications
  • Memory recalls maximum/minimum temperatures
  • Reading updates every second
Stainless steel probe (L×Ø: 203×3,5 mm).
Delivery information: Supplied with probe and cover, silver-oxide battery and Traceable® certificate of calibration.