VWR®, Environmentally Friendly Thermometers, Double Safe
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
620-0797EA 40.4 GBP
620-0797 620-0798 620-0792 620-0793 620-0794 620-0795
VWR®, Environmentally Friendly Thermometers, Double Safe
Thermometers Analogue Thermometers
These thermometers are easy to read quickly and accurately due to the high contrast black numbering against a yellow background. The numbers are printed larger than on a standard thermometer. Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
  • Enviro safe liquid
  • Non toxic, non hazardous, liquid
  • PFA coating prevents any contamination of liquid or glass
Traceable to NIST/DKD.
Certifications: Traceable toMore Product Information