Fold-back thermometer PT1000, TLC 1598
Supplier: EBRO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1340-1620
620-0226EA 174 GBP
620-0226
Fold-back thermometer PT1000, TLC 1598
Thermometers Digital Thermometers
This thermometer with foldable high accuracy PT1000 penetration sensor can be used for fast, accurate temperature monitoring in incoming goods inspections, catering, food products industry, trade and cool storage.
  • Robust and impact resistant
  • Short response time