Micro-Ubbelohde viscometers
Supplier: SI Analytics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
285401017 285401222 285401025 285401033 285401128 285401136 285401214 285401239 285401247 285401009 285401206 285401144 285401041
645-0071EA 495 GBP
645-0071 645-0077 645-0072 645-0073 616-3358 616-3359 645-0076 645-0078 645-0079 645-0070 645-0075 616-3360 645-0074
Micro-Ubbelohde viscometers
Viscometers Capillary Viscometers
Viscometers, with suspended ball level, for determination of relative or absolute kinematic viscosity.
  • Suitable for small samples and short running times
  • Ring marks for checking by manually
  • Uncalibrated or calibrated with constant for automatic measurements
Certifications: DIN 51 562-2