Matrix™ Reagent Reservoirs
Supplier: THERMO MATRIX TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8086 8095 8096 8093 8094 8085 8075
MATR8086EA 121 GBP
MATR8086 613-7452 MATR8096 613-7453 613-7451 613-7450 613-5831
Matrix™ Reagent Reservoirs
Reagent Reservoirs
PP or PS. These reservoirs offer an extra-wide base to add rigidity and stability, helping to avoid spills.
  • 75 ml reservoir made of PP is autoclavable and reusable
  • Trough within a trough maximises the amount of liquid accessible to pipette tips when using small amounts of reagent
  • Pour-off spouts on all four corners reduces spillage when pouring reagents out of the reservoir
  • Graduations on inside wall enable quick measurement of remaining liquid
  • Sterile reservoirs are packaged in snap and tear bags so packaging maintains sterility while being easy to open
More Product Information
Limited Time Sale
Select items below are currently on sale
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact Low Manufacturing Impact
Environmental certification IRECS; ISO 14001 (Certificate Registration No. EMS 752603) NAR Certificate of Retirement