VWR®, Single Channel Pipettes, Mechanical, Fixed Volume
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
613-0172EA 175 GBP
613-0172 613-7119 613-7120 613-7121 613-7122 613-7123 613-7124 613-7125 613-7126 613-7127 613-7128
VWR®, Single Channel Pipettes, Mechanical, Fixed Volume
Pipettes Micropipettes
Ergonomically designed to fit large or small hands for extra comfort while working, for left or right handed operation.
  • Slim tip mounting cone compatible with all tips
  • Stainless steel movable and height-adjustable tip ejector
  • Maintenance-free seals, polished plungers for superior precision
  • Fully autoclavable
Ordering information: Supplied with a key for laboratory calibration.