MyPIPETMAN® Select Pipettors, Gilson
Supplier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FP10012S FP10014S FP10001S FP10002S FP10003S FP10004S FP10006S FP10007S FP10008S FP10009S FP10010S FP10011S FP10013S FP10015S FP10016S
613-7020EA 1027 GBP
613-7020 613-7022 613-7009 613-7010 613-7011 613-7012 613-7014 613-7015 613-7016 613-7017 613-7018 613-7019 613-7021 613-7023 613-7024
MyPIPETMAN® Select Pipettors, Gilson
Pipettes Micropipettes
Ergonomic, safe, and reliable, MyPIPETMAN® Select offers volume setting choice and optional handle personalisation for optimal use in various protocols.
  • Free volume setting position for quick and effortless volume adjustment
  • Light ratchet volume setting for precise volume adjustment
  • Volume locked position to secure pipetting
Ergonomic design
Lightweight and well-balanced, MyPIPETMAN® Select has an elegant design with a textured effect for a better grip. With no compromise on quality and performance, the built-in ejection system with an integrated leverMore Product Information