Stands for HandyStep® touch pipettes
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
705220 705230 705235
613-6206EA 263 GBP
613-6206 613-6208 613-6209
Stands for HandyStep® touch pipettes
Pipette Stands
These stands and the universal holder are for use with HandyStep® touch und HandyStep® touch S pipettes.
The holding stand for the HandyStep® securely holds the device, even with a PD-Tip II inserted, and it saves space on the laboratory bench.
The charging stand for the HandyStep® touch saves space on the laboratory bench and securely holds the device even with a PD-Tip II attached. It features inductive charging technology, which enables charging without cable clutter. Simply place the HandyStep® touch into the stand, and the charging process begins.