Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Supplier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F81059 F81053 F81048 F81051 F81054 F81055 F81056 F81057 F81052 F81049 F81050 F81058
613-6058EA 1098 GBP
613-6058 613-6052 613-6047 613-6050 613-6053 613-6054 613-6055 613-6056 613-6051 613-6048 613-6049 613-6057
Multi-channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Pipettes Micropipettes
PIPETMAN® M Connected is a Bluetooth®-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes. Requiring virtually no pipetting forces to aspirate and dispense samples, PIPETMAN M Connected helps reduce pipetting fatigue and increase pipetting efficiency. The pipette interacts in real-time with TRACKMAN® Connected making planning and executing pipetting protocols faster, easier, and more reliable.
  • Simple and intuitive motorized pipette
  • Ultimate consistency
  • Connect to Gilson Apps
With PipetteScope®,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable