Single channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Supplier: GILSON
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
F81044 F81045 F81046 F81043 F81040 F81047 F81041 F81042
613-6043EA 507 GBP
613-6043 613-6044 613-6045 613-6042 613-6039 613-6046 613-6040 613-6041
Single channel pipettes, electronic, variable volume, PIPETMAN® M Connected
Pipettes Micropipettes
PIPETMAN® M Connected is a Bluetooth®-enabled electronic pipette that offers accuracy and precision in both standard and repetitive pipetting modes. Requiring virtually no pipetting forces to aspirate and dispense samples, PIPETMAN M Connected helps reduce pipetting fatigue and increase pipetting efficiency. The pipette interacts in real-time with TRACKMAN® Connected making planning and executing pipetting protocols faster, easier, and more reliable.
  • Simple and intuitive motorized pipette
  • Ultimate consistency
  • Connect to Gilson Apps
With PipetteScope®,More Product Information