Single-channel pipettes, mechanical, variable volume, Tacta
Supplier: Sartorius
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LH-729010 LH-729020 LH-729030 LH-729050 LH-729060 LH-729070 LH-729080 LH-729090
613-5478EA 318 GBP
613-5478 613-5479 613-5480 613-5481 613-5482 613-5483 613-5484 613-5485
Single-channel pipettes, mechanical, variable volume, Tacta
Pipettes Micropipettes
This mechanical pipette, with large display, has been designed with comfort in mind. Tacta makes pipetting effortless and safe, while producing accurate and reliable results. The pipette is made from carefully selected materials, with each component designed to meet the highest standards.
  • Comfortable to hold due to the ergonomically designed handle
  • Low pipetting and tip ejection forces that reduce the risk of Work Related Upper Limb Disorder (WRULD)
  • Unique Sartorius Optilock system provides flexibility for volume adjustment and locking
  • Controlled and
More Product Information