VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
613-5258EA 262 GBP
613-5258 613-5259 613-5260 613-5267 613-7129 613-7130 613-7131 613-7132 613-7133 613-7134 613-7135 613-7136 613-7137 613-7138 613-7139 613-7140 613-7141
VWR® Ergonomic High Performance (EHP), Single Channel Pipettes, Mechanical, Variable Volume
Pipettes Micropipettes
Ergonomic high performance pipettes provide accuracy and precision you can rely on. The upgraded, ergonomic features deliver comfortable pipetting, even during extensive periods of time.
  • Minimised pipetting and tip ejection forces
  • Effortless volume adjustment with volume lock
  • Superior accuracy and precision for critical applications
  • Tapered shaft and adjustable ‘Easy Grip’ ejector for universal tip fit
  • Extended finger rest for convenient pipetting
  • UV-resistant and fully autoclavable