Serum collection tubes, vacuum, BD Vacutainer®
Supplier: BD Medical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
367624 369032 368492 368815 367614 367896 367837
613-3974EA 103 GBP
613-3974 BDAM369032 BDAM368492 BDAM368815 BDAM367614 BDAM367896 BDAM367837
Serum collection tubes, vacuum, BD Vacutainer®
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
PET, transparent.
  • With or without additive
  • With label
In order to obtain serum samples from plastic tubes, the tube must have a coagulation activator added. As the plastic surface alone is insufficient to trigger the coagulation within an acceptable time, BD Vacutainer® Plus plastic serum tubes have silica particles added for this purpose. These tubes are marked with the acronym CAT (Clot Activator Tube).