Digital burettes, Titrette®
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4760151 4760161 4760251 4760261 4760451 4760141 4760241
613-0795EA 986 GBP
613-0795 613-0796 613-0905 613-0906 613-3840 613-2314 613-5796
Digital burettes, Titrette®
Burettes
Variable volume.
  • Light, flexible burette with compact design and high accuracy
  • Smooth running operation with large, easy grip hand wheels for sensitive, dropwise titration
  • Easier use: Adjustment with easy calibration, calibration schedule to save a date for the next calibration, save power with Automatic Off function, change decimal place settings
  • Easy to dismantle: Cleaning, maintenance and part replacement
  • PC interface (optional) - for data transmission directly to a PC, which eliminates transmission errors during transcription of the primary data to
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable Low Manufacturing Impact,Sustainable Packaging
Environmental certification ISO 14001 and ISO 50001