Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Supplier: EPPENDORF
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0030089.456 0030089.480 0030089.464 0030089.448 0030089.430 0030089.421 0030089.413 0030089.405 0030089.472 0030089.936
613-2063EA 124 GBP
613-2063 613-2066 613-2064 613-2062 613-2061 613-2060 613-2059 613-2058 613-2065 613-3857
Combitips® advanced Eppendorf Quality Positive Displacement Pipette Tips
Pipette Tips
Eppendorf Combitips® advanced, in 'Eppendorf Quality' grade, offer precise, positive displacement pipetting. Ideal for handling challenging liquids, ensuring accuracy and efficiency in diverse lab applications with their versatile and durable design.
  • Combitips® advanced tips are the dedicated consumable for all Repeater®/Multipette® multi-dispenser pipettes from Eppendorf
  • 9 volume sizes (0,1 to 50 ml) offer a maximum range of dispensing volumes, with increments as low as 100 nanoliters
  • Combitips® advanced in ‘Eppendorf Quality’ are reliable, easy-to-handle
More Product Information