Methylene blue 10 g/l in aqueous solution

Supplier: VWR Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

6129.1000EA 78.4 GBP
6129.1000 6129.9010
Methylene blue 10 g/l in aqueous solution
Methylene blue
Formula: C₁₆H₁₈ClN₃S
Density: 1.01 g/cm³ (20 °C)
MDL Number: MFCD00150008
CAS Number: 61-73-4
EINECS: 200-515-2

Order Now

Specification Test Results

Identification Passes test
Control weight QC checked


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR