VWR® Burettes, with PTFE Key Stopcock
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-6691EA 73.8 GBP
612-6691 612-6692 612-6693
VWR® Burettes, with PTFE Key Stopcock
Burettes
Borosilicate 3.3 glass, class AS.
  • High chemical and thermal resistance
  • Schellbach stripe for accurate meniscus reading
  • Stopcock with PTFE key
  • High contrast blue enamel graduations
  • Batch certificate included
To avoid material damaging friction between the burette and the stopcock and to ensure tightness, joint grease is necessary when working with burettes that have a glass stopcock. After use the joint grease must be removed as the burette tip could become contaminated. This step is not necessary when working with burettes with a PTFE key stopcock.
Order Now
SPECIFICATIONS
Environmentally Preferable