VWR®, Mixing Measuring Cylinders, Tall Form, Class A
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-5771EA 40.4 GBP
612-5771 612-5772 612-5773 612-5774 612-5775 612-5776 612-5777
VWR®, Mixing Measuring Cylinders, Tall Form, Class A
Cylinders Measuring Cylinders
Borosilicate 3.3 glass, class A.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, NS ground and PE stopper
  • Ring marks at major graduations, marks and inscriptions in blue enamel
Certifications: DIN EN ISO 4788
Ordering information: Batch certificate included.