VWR®, Measuring Cylinders, Tall Form, Class B, PP
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-5602EA 2.1 GBP
612-5602 612-5604 612-5639 612-5658 612-5659 612-5665 612-5666 612-5667
VWR®, Measuring Cylinders, Tall Form, Class B, PP
Cylinders Measuring Cylinders
PP, translucent, tall form, class B.
  • Large hexagonal base for better stability
  • Graduations are calibrated in mold for accuracy and consistency
  • Withstand temperatures of 0 to 121°C
  • Very good chemical resistance
  • Resin meets US FDA 21 CFR requirements
Certifications: DIN 12681/ISO 6706