Bulb pipettes, one mark, class AS, brown graduation
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1340129 1340133 1340139 1340140 1340145 1340148 1340150 1340155 1340156 1340157 1340158 1340159 1340160 1340164 1340165 1340170 1340173 1340174 1340175 1340180 134012908 134013308 134014008 134014508 134014808 134015008 134015508 134015608 134015708 134015808 134015908 134016008 134016408 134016508 134017008 134017308 134017408 134017508 134018008 134013908 134013327 134015527 134016027 134017027 134017527 134018027 1340165DKD 134014027 134014827 134015027 134015627 134015727 134015827 134015927 134016427
612-1321EA 11.9 GBP
612-1321 612-1322 612-1323 612-1324 612-1325 612-1326 612-1327 612-1328 612-1329 612-1330 612-1331 612-1332 612-1333 612-1334 612-1335 612-1336 612-1337 613-3217 612-1339 612-1340 612-5753 HIRS134013308 HIRS134014008 HIRS134014508 HIRS134014808 HIRS134015008 HIRS134015508 HIRS134015608 HIRS134015708 HIRS134015808 HIRS134015908 HIRS134016008 HIRS134016408 HIRS134016508 HIRS134017008 HIRS134017308 HIRS134017408 HIRS134017508 HIRS134018008 HIRS134013908 612-5454 612-5458 612-5463 612-5466 612-5467 612-5468 612-5465 612-5455 612-5456 612-5457 612-5459 612-5460 612-5461 612-5462 612-5464
Bulb pipettes, one mark, class AS, brown graduation
Pipettes Bulb Pipettes
AR-Glas®.
  • Calibrated to deliver (TD, Ex)
  • With silver diffusion ink durable brown graduation
  • Available with different certificates
DIN EN ISO 648
Certifications: DIN EN ISO 648; DE-M marking