Mixing measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
32462 32438 32448 32408 32420 32428 32454 932408
612-5029EA 183 GBP
612-5029 612-5026 612-5027 612-5023 612-5024 612-5025 612-5028 BRND932408
Mixing measuring cylinders, tall form, class A, borosilicate glass, BLAUBRAND®
Cylinders Measuring Cylinders
Borosilicate glass 3.3, class A, conformity certified.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base, PP stopper (except for 612-5028 and 612-5029 octagonal PE stopper)
  • Ring marks at major graduations, marks and inscriptions in high contrast blue enamel
  • Total order from one batch
* Height without stopper
Certifications: DIN EN ISO 4788.
Ordering information: Supplied with a batch certificate. Measuring cylinders are also available on request with an individual certificate, a USP individual certificate or a DAkkS calibration certificate.