Burettes, straight stopcock, PYREX®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3295/24ASWC 3295/22ASWC 3295/26ASWC 3295/28ASWC 3295/12AS 3295/14AS 3295/16AS 3295/18AS
612-3970EA 74.4 GBP
612-3970 612-3959 612-3971 612-3972 612-3973 612-3974 612-3975 612-3976
Burettes, straight stopcock, PYREX®
Burettes
Borosilicate glass with interchangeable PTFE key, Class AS.
  • Calibrated to deliver ‘Ex’
  • PTFE key for grease-free perfomance
  • White enamel graduations for good visibility with dark coloured solutions
Certifications: ISO 385.