Measuring cylinders, tall form, class A, PMP, Azlon®
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CTA0010P CTA0025P CTA0050P CTA0100P CTA0250P CTA0500P CTA1000P
612-3917EA 17.4 GBP
612-3917 612-3918 612-3919 612-3920 612-3921 612-3922 612-5696
Measuring cylinders, tall form, class A, PMP, Azlon®
Cylinders Measuring Cylinders
PMP (TPX®), transparent, tall form, class A.
  • Hexagonal base with non-slip, anti-suction feature, spout, excellent chemical resistance
  • Enhanced pin sharp blue printed graduations
  • Autoclavable - Does not affect accuracy
Certifications: ISO 6706