VWR® Smoothie™ Pipette Filler
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-3862EA 0 GBP
612-3862 612-3863 612-3864 612-3865
VWR® Smoothie™ Pipette Filler
Pipettes Pipette Controllers
The Smoothie™ pipette filler is a simple manual pipetting aid for convenient single-handed operations. It fits all sizes of glass or plastic pipettes from 1 to 25 ml.
  • Comfortable and simple to use
  • Smooth, controllable thumb action
  • Silicone adapter for leakproof positioning of each pipette
The unique range control allows selection from two dispense resolutions. One range setting delivers fast filling with optimal accuracy when used with 10 to 25 ml pipettes, the other setting is for use with pipettes from 1 to 10 ml where accuracy of fill can sometimesMore Product Information