VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-3837EA 68.4 GBP
612-3837 612-3840 612-3838 612-3836 612-3835 612-3834 612-3833 612-3832 612-3839
VWR® Measuring Cylinders, Tall Form, Class A, Borosilicate Glass
Cylinders Measuring Cylinders
Borosilicate 3.3 glass, class A, tall form.
  • Calibrated to contain (TC, In)
  • Hexagonal glass base with spout
  • Blue graduations with ring marks at major graduations
DIN 12680
Certifications: DIN 12680
Ordering information: Batch number and batch certificate included.