Volumetric flasks, class A, white graduation, works certified
Supplier: SciLabware
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3152/08A 3152/10A 3152/14A 3152/16A 3152/20A 3152/04A 3152/06A 3152/12A 3152/18A 3152/22A
612-3248EA 98.5 GBP
612-3248 612-3251 612-3244 612-3243 612-3247 612-2903 612-2904 612-3249 612-3242 612-3245
Volumetric flasks, class A, white graduation, works certified
Flasks Volumetric Flasks
PYREX®, borosilicate glass, amber, with PE stopper.
  • Calibrated to the tolerances required by the United States Pharmacopeia (USP), which are tighter than those of ISO/DIN standards
  • Each flask is laser etched with a unique individual serial number, batch code and date of manufacture - Batch certificates are downloadable from website: www.scilabware.com
  • White enamel inscriptions, ideal for use with light sensitive solutions
Certifications: BS EN ISO 1042, DIN 12664