Measuring cylinders, tall form, PPCO or PMP, economy, Nalgene®
Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3664-0010 3664-0025 3664-0050 3664-0100 3664-0250 3664-0500 3664-1000 3665-0010 3665-0025 3665-0050 3665-0100 3665-0250 3665-0500 3665-1000
NALG3664-0010EA 4 GBP
NALG3664-0010 612-2867 612-2868 612-0600 612-2869 612-2870 612-2871 612-2872 612-2873 612-2874 612-2875 612-2876 612-2877 612-2878
Measuring cylinders, tall form, PPCO or PMP, economy, Nalgene®
Cylinders Measuring Cylinders
Measuring cylinders, made of PPCO, translucent or PMP, clear, tall form.
  • Calibrated “To Contain/To Deliver"
  • No meniscus to confuse readings allow for accurate measurements
  • Easy to read molded graduations
  • Stable with large round base