Dissolved oxygen bottles
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
6500183 6500187
612-2794EA 65.2 GBP
612-2794 215-1245
Dissolved oxygen bottles
Bottles Dissolved Oxygen Bottles
Oxygen bottles according to Winkler, with white inscription label.
  • The adjusted capacity is individually marked on the bottle
  • A unique serial number is engraved on the bottle and stopper