VWR®, Spreaders, Triangle Shaped, Disposable
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-2687EA 90.6 GBP
612-2687 612-2688 612-2689 612-2690
VWR®, Spreaders, Triangle Shaped, Disposable
Spreaders Cell Spreaders
PP, sterile (individually wrapped) or non sterile (bulk packed). Convenient and economical, these cell spreaders are designed to simplify bacterial culture work. The foot of the spreader has a completely smooth, rounded surface, free of rough edges and imperfections. It enables even spreading of liquid samples across the surface of agar plates without gouging or cutting the medium.
  • Durable enough to be autoclaved and used again
  • Width: 30 mm (blue)/60 mm (yellow)