VWR®, Spreaders, T-shaped
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
612-2651EA 141 GBP
612-2651 612-2652 612-2653
VWR®, Spreaders, T-shaped
Spreaders Cell Spreaders
PS, sterile. Designed for spreading and dispersing liquids across the surface of agar culture plates. Used for performing bacterial (for example, CFU) plate counts on water, milk and other liquid samples.
  • Smooth, rounded surfaces to prevent cutting or gouging of agar
  • T-shaped to ensure even pressure is applied while spreading