Aspirating pipettes, Falcon®
Supplier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
357558 357501
734-0456EA 55.2 GBP
734-0456 612-2458
Aspirating pipettes, Falcon®
Pipettes Aspirating Pipettes
PS pipettes, non plugged, non graduated, disposable, sterile.
  • Safer alternative to glass Pasteur pipettes
  • Non pyrogenic
  • Individually packed to prevent contamination
Packaging: Individually packed in a thermo-formed paper/plastic wrap.