Pipette controller, Pi-Pump
Supplier: KARTELL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5050 5051 5052
612-1925EA 68.8 GBP
612-1925 612-1926 612-1927
Pipette controller, Pi-Pump
Pipettes Pipette Controllers
Pipetting aid for all plastic and glass pipettes.
  • Suction and dispensing achieved by rotating the wheel
  • Liquid evacuated by pressing the lever