Bulb pipettes, one mark, class B
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1300129 1300133 1300140 1300148 1300150 1300155 1300160 1300164 1300165 1300170 1300175 1300180
612-1348EA 11.6 GBP
612-1348 612-1349 612-1359 612-1350 612-3464 612-1351 612-1352 612-1353 612-1354 612-1355 612-1357 612-1358
Bulb pipettes, one mark, class B
Pipettes Bulb Pipettes
AR-Glas®
  • Calibrated to deliver (TD, Ex)
  • With silver diffusion ink durable brown graduations
DIN 12690
Certifications: DIN EN ISO 648; DE-M marking