Graduated pipettes, with piston
Supplier: HIRSCHMANN
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
1190133 1190141 1190153 1190160 1190165 1190170
612-1025EA 37.7 GBP
612-1025 612-1026 612-1027 612-1028 612-1029 612-1030
Graduated pipettes, with piston
Pipettes Graduated Pipettes
AR-Glas®.
  • Calibrated to deliver ‘Ex’
  • Durable amber stain graduation, main point ring graduation
  • Integrated syringe pipetting aid