Syringes, FORTUNA® OPTIMA®, Luer tip
Supplier: Poulten & Graf
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7.10221 7.10227 7.10229 7.10233 7.10237 7.10241 7.10244 7.10245 7.10251
612-0311EA 10.8 GBP
612-0311 612-0312 612-0313 612-0314 612-0315 612-0316 612-0317 612-0318 612-0319
Syringes, FORTUNA® OPTIMA®, Luer tip
Syringes Syringes, Medical
Barrel and piston in Durobax® glass, without needle.
  • Luer tip
  • Graduation in brown
  • Autoclavable at +134 °C
Certifications: Conform to ISO 9001
Ordering information: Supplied individually packaged.
Order Now
SPECIFICATIONS
Environmentally Preferable