Weighing bottles, glass
Supplier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05560002 05560005 05560008 05560012 05560015 05560019 05560022 05560025 05560028 05560502 05560504 05560509 05560513 05560516 05560521
611-9311EA 29.3 GBP
611-9311 611-9312 611-9313 611-1234 611-9317 611-1235 611-9314 611-9315 611-9316 611-9302 611-9303 611-9304 611-9305 611-9306 611-9307
Weighing bottles, glass
Bottles Weighing Bottles
DURAN® borosilicate glass 3.3.
  • Interchangeable lids with ground joint
  • Writing area in white
Order Now
SPECIFICATIONS
Environmentally Preferable