VWR®, Weighing Boats
Supplier: VWR Collection
Ratings:
Avg. Ratings: 5.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
611-9194EA 63.4 GBP
611-9194 611-9195 611-9196
VWR®, Weighing Boats
Boats Weighing Boats
Borosilicate 3.3 glass.
  • Ideal for weighing small samples
  • Powders can be tapped or rinsed through the tubular stem of the weighing boats into a receiving vessel
Order Now
SPECIFICATIONS
Environmentally Preferable