VWR®, Calibration Weight Sets, Class F2
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-8662EA 249 GBP
611-8662 611-8663 611-8664 611-8665 611-8666 611-8667 611-8668 611-8669 611-8671 611-8672 611-8673 611-8674 611-8675 611-8676 611-8677 611-8679 611-8680 611-8681 611-8682 611-8692 611-8693 611-8694 611-8695 611-8696 611-8697 611-8698 611-8699 611-8701 611-8702 611-8703 611-8704 611-8705 611-8706 611-8707 611-8709 611-8710 611-8711 611-8712 611-4936 611-4937 611-4942 611-4943
VWR®, Calibration Weight Sets, Class F2
Weights Calibration Weights
Stainless steel. Wire weights up to 500 mg in cobalt alloy.
  • Identification number guarantees traceability for weights from 1 g upwards
  • Laser engraving improves legibility of the markings
  • Available with or without COFRAC certificate
Certifications: Conform to OILM R111 recommendations.
Order Now
SPECIFICATIONS
Environmentally Preferable