VWR® Premium Precision Balances
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-4801EA 896.84 GBP
611-4801 611-4802 611-4803 611-4804 611-4805 611-4806 611-4807 611-4808 611-4809 611-4810 611-4811 611-4812 611-4813 611-4814 611-4815 611-4816 611-4817 611-4818 611-4819 611-4820 611-4821 611-4822 611-4823 611-4824 611-4825 611-4826 611-4827 611-4828 611-4829 611-4830 611-4831 611-4832 611-4833 611-4834 611-4835 611-4836
VWR® Premium Precision Balances
Balances and Scales
A selection of 36 high level balances offering high accuracy and repeatability combined with easy, intuitive operation and a large touch screen for applications in the laboratory, industrial and educational settings.
  • Highly accurate results (high-end weighing technology)
  • Fast stabilisation times
  • Easy to clean
  • High chemical resistance
  • Control panel with high resolution, touch screen functionality and eight language options
  • Overload protection
  • Future-proof USB type C interface
  • Under bench weighing
  • Data transfer as PC spreadsheet, PC text and Ascii protocols
More Product Information