Portable balances, Navigator NV and NVT series
Supplier: OHAUS
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
30456382 30456383 30456384 30456385 30456386 30456387 30456388 30456389 30456390 30456391 30456392 30456393 30456394 30456395 30467835 30467836 30467838 30467837 30615581 30615582 30615583 30615584 30615585 30615586 30615587
611-4459EA 273 GBP
611-4459 611-4460 611-4464 611-4465 611-4466 611-4467 611-4468 611-4469 611-4470 611-4478 611-4479 611-4480 611-4484 611-4485 611-4666 611-4667 611-4669 611-4668 611-5054 611-5055 611-5056 611-5057 611-5058 611-5059 611-5060
Portable balances, Navigator NV and NVT series
Balances and Scales
Navigator portable balances are perfect for a diverse range of weighing applications in a laboratory or industrial and education settings. Navigator is a powerful balance that is rugged and multifunctional, has fast stabilisation time, overload protection at four times its rated capacity, and is easy to operate.
  • Designed for accuracy and repeatability in essential weighing applications
  • Improved productivity and throughput with fast stabilisation time (less than one second)
  • NV models have been designed to conveniently stack, helping to save storage space
More Product Information