VWR® Professional TP / TPW, Precision Balances with Touch Screen
Supplier: VWR Collection
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-3692EA 1350 GBP
611-3692 611-3693 611-3688 611-3689 611-3690 611-3694 611-3700 611-3701 611-3695 611-3696 611-3697 611-3703 611-3698 611-3699 611-3704 611-3706 611-3826 611-3827 611-3858 611-3857 611-3861 611-3862 611-4677 611-4678
VWR® Professional TP / TPW, Precision Balances with Touch Screen
Balances and Scales
VWR® Professional precision balances are ideal for wide range of laboratory weighing tasks, they have been designed for multi-user environments (each user profile has password protection). These interactive balances, which feature an internal database to store data and drying methods, help improve efficiency and productivity. All professional models have a 5" touch screen with icon-driven menu, qwerty touch keypad and a stainless steel top pan. Models are GLP set-up, storing balance, project and user identification data, as well as date and time. GLP information can be also printed out withMore Product Information