High capacity balances, GF-K and GX-K series
Supplier: AnD Instruments Ltd (A&D)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
GF-8K2 GF-12K GF-20K GF-10K GF-30K GX-32K GX-8K2
611-3082EA 0 GBP
611-3082 611-3076 611-3077 611-3075 611-3078 611-3106 611-3108
High capacity balances, GF-K and GX-K series
Balances and Scales
High capacity top loading balances from A&D featuring high speed weighing and settling times. Applications include counting, percentage weighing, animal weighing, below balance weighing and density determination. All models feature display auto-off function, overload protection, standard comparator function (HI/OK/LO annunciator) and counting function with Automatic Counting Accuracy Improvement (ACAI). RS232C interface for printer or computer for GLP data output format to suit quality systems.
  • Rugged aluminium alloy enclosure and stainless steel pan with IP 65 waterproof and dust
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable