Precision balances, EK-i series
Supplier: AnD Instruments Ltd (A&D)
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EK-4100I EK-410I EK-1200I EK-12KI EK-2000I EK-200I EK-3000I EK-300I EK-6100I-EC EK-610I-EC EK-6000I
611-3018EA 0 GBP
611-3018 611-3019 611-2976 611-2977 611-2978 611-2979 611-3016 611-3017 611-3022 611-3023 611-3020
Precision balances, EK-i series
Balances and Scales
Compact, lightweight, low profile balances from A&D with a rugged housing and stainless steel pan for reliable, accurate weighing. External calibration with full digital calibration and user definable mass. The sealed key panel protects against dust and spills. All models feature auto-off function, overload protection, standard comparator function (HI/OK/LO annunciator) and counting function with Automatic Counting Accuracy Improvement (ACAI). GLP conformity with ID and serial number outputs.
  • Nine built-in weighing units plus counting and percentage software
  • Large backlit LCD
More Product Information